Learn More
LMOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151171AP
Catalog No. 50-172-9994
Add to cart
Description
Description
The leiomodin 1 protein (LMOD1) has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves ft. disease and thyroid-associated ophthalmopathy. The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves ft. disease and thyroid-associated ophthalmopathy.
Specifications
Specifications
LMOD1
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0A0G2K0D3, P29536, Q8BVA4
1D, 64 kDa autoantigen 1D, 64 kDa autoantigen 1D3, 64 kDa autoantigen D1, 64kD, D1, Leiomodin 1, leiomodin 1 (smooth muscle), Leiomodin, muscle form, LMOD1, SM LMOD, Smooth muscle leiomodin
Rabbit
IgG
150 μL
-20°C
Primary
25802, 304816, 93689
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
LMOD1
Liquid
Lmod1
LMOD1 Fusion Protein Ag7176
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat