Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMP2/PSMB9 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI1D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271261APCCY7

Catalog No. N271261APC7


Add to cart

Description

Description

LMP2/PSMB9 Monoclonal antibody specifically detects LMP2/PSMB9 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

LMP2/PSMB9
OTI1D1
Flow Cytometry
Affinity Purified, Conjugated
PSMB9
Recombinant protein fragment corresponding to amino acids 21-219 of human LMP2/PSMB9 (NP_002791) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), Really interesting new gene 12 protein, RING12EC 3.4.25.1
Mouse
IgG2a
0.1 mL
Primary
5698.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit