Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMX1b Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LMX1b
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093783
View Documents
Novus Biologicals
NBP304700100UL
100 μg Each for $320.71
Add to cart
 
NB093784
View Documents
Novus Biologicals
NBP30470020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

LMX1b Polyclonal antibody specifically detects LMX1b in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

LMX1b
Unconjugated
Rabbit
Neuroscience
PBS with 50% glycerol, pH7.3.
4010
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Western Blot
Purified
RUO
Human, Mouse
LIM homeobox transcription factor 1, beta, LIM homeobox transcription factor 1-beta, LIM/homeobox protein 1.2, LIM/homeobox protein LMX1B, LMX1.2, LMX-1.2, MGC138325, MGC142051, NPS1
Recombinant fusion protein containing a sequence corresponding to amino acids 136-395 of human LMX1B (NP_002307.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit