Thermo Scientific™ LN2 Level Monitor
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
Manufacturer: Thermo Scientific™ CN509X7
 View more versions of this product
Catalog No. 13-762-363
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Locator 4 Plus
    Type LN2 Measuring Device
    Voltage 230V,230V
    Description Level Monitor
    For Use With Locator 8
    Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
    Ultrasonic Level Monitor
    • Minimizes conduction and liquid nitrogen evaporation
    • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
    • Audible and visual alarms signal low level liquid nitrogen conditions
  • Description & Specifications
Related Products
Product Content Feedback