Learn More
Thermo Scientific™ LN2 Level Monitor
Safeguard irreplaceable biological specimens with the Thermo Scientific? LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
Manufacturer:  Thermo Scientific™ CN509X770
 View more versions of this product
Catalog No. 13-762-366
  Check Availability
Add to cart
Specifications
Description
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
  • Minimizes conduction and liquid nitrogen evaporation
  • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
  • Audible and visual alarms signal low level liquid nitrogen conditions
Description & Specifications
Specifications
Level Monitor
LN2 Measuring Device
Locator 8 Plus
Locator 8 Plus
100-240V