Learn More
Thermo Scientific™ LN2 Level Monitor
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
$2080.00 - $2110.00
Specifications
Description Level Monitor
View More Specs
Products 2
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
13-762-363
View Documents
Thermo Scientific™
CN509X7
230 V Each for $2,110.00
Add to cart
13-762-366
View Documents
Thermo Scientific™
CN509X770
100 - 240 V Each for $2,080.00
Add to cart
Description
Description
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
  • Minimizes conduction and liquid nitrogen evaporation
  • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
  • Audible and visual alarms signal low level liquid nitrogen conditions
  • Warranty varies by country, please contact us for details
Specifications
Specifications
Level Monitor