Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
LNK/SH2B3 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF5888SP
Catalog No. AF5888SP
This item is not returnable. View return policy
Description
Description
LNK/SH2B3 Polyclonal antibody AF5888SP specifically detects LNK/SH2B3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
LNK/SH2B3
LYOPH
Western Blot 0.5 ug/mL
Q9UQQ2
IDDM20, LNKSH2B adapter protein 3, Lymphocyte adapter protein, Lymphocyte-specific adapter protein Lnk, SH2B adaptor protein 3, SH2B3, Signal transduction protein Lnk
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human LNK/SH2B3 in Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SH2B3
E. coli-derived recombinant human LNK/SH2B3 Gln427-Leu575 Accession # Q9UQQ2
Immunogen affinity purified
RUO
Primary
10019
Reconstitute at 0.2 mg/mL in sterile PBS.