Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LNK Mouse anti-Human, Clone: OTI2B11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525516

Catalog No. PIMA525516


Add to cart

Description

Description

APS (adapter molecule containing PH and SH2 domains), SH2-B and Lnk compose a family of adapter proteins, which contain a pleckstrin homology (PH) domain, an SH2 domain and a tyrosine phosphorylation site. Stimulation of B cell receptor (BCR) or T cell receptor (TCR) results in the phosphorylation of the immunoreceptor tyrosine-based activation motif (ITAM) of BCR, TCR and several substrates. APS, SH2-B and Lnk may bind to the ITAM domain of BCR and TCR. Lnk is tyrosine phosphorylated in response to TCR stimulation and APS has been shown to be tyrosine phosphorylated in response to BCR stimulation.
Specifications

Specifications

LNK
Monoclonal
0.61 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UQQ2
SH2B adapter protein 3; Lymphocyte adapter protein; Lymphocyte-specific adapter protein Lnk; Signal transduction protein Lnk; SH2B3; LNK
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10019
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B11
Unconjugated
SH2B3
Liquid
SH2B3
Full length human recombit protein of SH2B3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit