Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Lobeline hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
134-63-4
C22H28ClNO2
373.921
MKMYPTLXLWOUSO-OCIDDWSYSA-N
--lobeline hydrochloride
124081008
2-[(2S,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;hydrochloride
CN1C(CCCC1CC(=O)C2=CC=CC=C2)CC(C3=CC=CC=C3)O.Cl
This item is not returnable. View return policy
134-63-4
C22H28ClNO2
373.921
MKMYPTLXLWOUSO-OCIDDWSYSA-N
--lobeline hydrochloride
124081008
2-[(2S,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;hydrochloride
CN1C(CCCC1CC(=O)C2=CC=CC=C2)CC(C3=CC=CC=C3)O.Cl

Nicotinic partial agonist

High affinity, partial agonist with a Ki value of 4.4 nM in rat brain.
Quantity 100mg
Formula Weight 373.92
Percent Purity >98%
Chemical Name or Material (-)-Lobeline hydrochloride

Recommended Storage : Store at Room Temperature