Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRABD2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548267

Catalog No. PA548267

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Metalloprotease that acts as a negative regulator of the Wnt signaling pathway by mediating the cleavage of the 8 N- terminal residues of a subset of Wnt proteins. Following cleavage, Wnt proteins become oxidized and form large disulfide-bond oligomers, leading to their inactivation. Able to cleave WNT3A, WNT5, but not WNT11. Required for head formation.
Specifications

Specifications

TRABD2A
Polyclonal
PBS with 0.09% sodium azide
Q86V40
Metalloprotease TIKI1, 34--, TRAB domain-containing protein 2A, TRABD2A, C2orf89, TIKI1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
129293
Western Blot
Unconjugated
TRABD2A
Liquid
TRABD2A
KLH conjugated synthetic peptide between 342-371 amino acids from the C-terminal region of human LOC129293.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit