Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ LOC130940 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00130940T01
Catalog No. 89-950-830
Description
Description
Specifications
Specifications
293T
BC015395
pCMV-LOC130940 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LOC130940 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 30.3kDa
Transient overexpression cell lysate was tested with anti-LOC130940 antibody (Mfr. No. H00130940-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human