Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C19orf84 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548305

Catalog No. PA548305

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

C19orf84
Polyclonal
PBS with 0.09% sodium azide
I3L1E1
hypothetical protein LOC147646
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
147646
Western Blot
Unconjugated
C19orf84
Liquid
C19ORF84
KLH conjugated synthetic peptide between 147-176 amino acids from the C-terminal region of human LOC147646.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit