Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC284009 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen LOC284009
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170605
View Documents
Novus Biologicals
NBP170605
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

LOC284009 Polyclonal specifically detects LOC284009 in Human samples. It is validated for Western Blot.
Specifications

Specifications

LOC284009
Polyclonal
Rabbit
Human
hypothetical LOC284009, MGC138239
LOC284009
IgG
Affinity Purified
Western Blot
Unconjugated
RUO
PBS and 2% Sucrose with 0.09% Sodium Azide
284009
Synthetic peptides corresponding to LOC284009(hypothetical protein LOC284009) The peptide sequence was selected from the N terminal of LOC284009. Peptide sequence IRCGRPAVVHIGGEGARWEKGARGRKEHRLRRSDLGSRPVPFLAQGIPDI.
Primary
Store at -20C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit