Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOC285033 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | LOC285033 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156378
|
Novus Biologicals
NBP156378 |
100 μL |
Each of 1 for $436.00
|
|
Description
LOC285033 Polyclonal specifically detects LOC285033 in Human samples. It is validated for Western Blot.Specifications
LOC285033 | |
Polyclonal | |
Rabbit | |
Human | |
Q3KRF4 | |
285033 | |
Synthetic peptides corresponding to LOC285033(hypothetical protein LOC285033) The peptide sequence was selected from the N terminal of LOC285033. Peptide sequence VIKEGAVCGIARPKTSRVNSSQDQIQVASENTHSGSLHQRPASGARLPAS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
hypothetical protein LOC285033, MGC126779, MGC126781 | |
LOC285033 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title