Learn More
Aviva Systems Biology Corp LOC339047 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP56035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC339047. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC339047. Alternate names: KIAA0220ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61701
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.