Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC342293 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191567

 View more versions of this product

Catalog No. NBP191567


Add to cart

Description

Description

LOC342293 Polyclonal antibody specifically detects LOC342293 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

LOC342293
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
LOC342293 ATP-binding cassette, sub-family A (ABC1), member 3 pseudogene
Rabbit
IgG
100ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Mouse: 100%; Crab-eating macaque: 100%; Caenorhabditis briggsae: 90%; Caenorhabditis vulgaris: 90%; Caenorhabditis elegans: 90%; Olive fruit fly: 84%; Bovine: 84%; Yellowfever mosquito: 84%; Tetrahymena thermophila SB210: 84%; Green puffer: 84%; Western clawed frog: 84%; Purple sea urchin: 84%; Canine: 84%; Chicken: 84%; Rat: 84%; Florida lancelet: 84%; Black-legged tick: 84%; Human: 100%; Trichoplax reptans: 84%; Choanoflagellate: 84%; Starlet sea anemone: 84%; Zebrafish: 84%.
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Purified
LOC342293
Synthetic peptide directed towards the N terminal of human LOC342293. Peptide sequence PQIGLYVNKCLNSLELESHAARLINTYSEGNKRRLSTAIALMGRSSVIFL.
Protein A purified
RUO
Primary
342293
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit