Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody (Nalpy3-b) - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$461.00
Specifications
Antigen | LOC342293 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191567
|
Novus Biologicals
NBP191567 |
100 μL |
Each of 1 for $461.00
|
|
|||||
Description
LOC342293 Polyclonal specifically detects LOC342293 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LOC342293 | |
Polyclonal | |
Purified | |
RUO | |
LOC342293 ATP-binding cassette, sub-family A (ABC1), member 3 pseudogene | |
LOC342293 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
342293 | |
Synthetic peptide directed towards the N terminal of human LOC342293. Peptide sequence PQIGLYVNKCLNSLELESHAARLINTYSEGNKRRLSTAIALMGRSSVIFL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title