Learn More
Aviva Systems Biology Corp LOC344065 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP51006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC344065. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC344065. Alternate names: -ApplicationWB. Species reactivity: Dog
Catalog No. 50-014-60482
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.