Learn More
Aviva Systems Biology Corp ACCSL Antibody - N-terminal region (ARP48528_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48528_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC390110. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC390110. Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59850