Learn More
Aviva Systems Biology Corp LOC390738 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP51011_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC390738. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC390738. Alternate names: -ApplicationWB. Species reactivity: Dog, Rat
Catalog No. 50-014-60484
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.