Learn More
Aviva Systems Biology Corp LOC391524 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP50964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC391524. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC391524. Alternate names: THRAP3LApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60474
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.