Learn More
Aviva Systems Biology Corp C17orf97 Antibody - N-terminal region (ARP54431_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54431_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC400566. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC400566. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61205