Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC400856 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191601

 View more versions of this product

Catalog No. NBP191601

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LOC400856 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

LOC400856
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
LOC400856
Synthetic peptide directed towards the C terminal of human LOC400856. Peptide sequence SLHTSPRAEGAGSGLSQPQRRALTAQGGAEGLLERGQSGHGGRGGAKSER.
Immunogen affinity purified
RUO
Primary
400856
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
XP_375929
hypothetical gene supported by AK123815
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit