Learn More
Aviva Systems Biology Corp WDPCP Antibody - N-terminal region (ARP56279_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC51057. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC51057. Alternate names: FRTZ, BBS15, FRITZ, C2orf86ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61794