Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ LOC619207 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Supplier: Abnova™ H00619207T01
Specifications
Western Blot | |
BC038300.1 | |
pCMV-LOC619207 full-length | |
100 μL | |
Human |
LOC619207 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 25.63kDa | |
Transient overexpression cell lysate was tested with anti-LOC619207 antibody (Mfr. No. H00619207-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction