Learn More
Aviva Systems Biology Corp LOC641765 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP51066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC641765. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC641765. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60492
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.