Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOC641765 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191408

 View more versions of this product

Catalog No. NBP191408

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LOC641765 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

LOC641765
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
LOC641765
Synthetic peptide directed towards the C terminal of human LOC641765. Peptide sequence SQNLERILAMANKITFTVTRPFQGLIRKPDEDDTNRLGEVILQEQVKELF.
66 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
XP_940624
similar to GTF2I repeat domain containing 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
641765
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit