Learn More
Aviva Systems Biology Corp LOC652618 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP49306_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC652618. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC652618. Alternate names: -ApplicationWB. Species reactivity: Pig, Rat
Catalog No. 50-014-60057
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.