Learn More
Abnova™ LOC653107 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00653145T01
Catalog No. 89-935-169
Add to cart
Description
Description
Specifications
Specifications
293T
NM_001040084.1
pCMV-LOC653107 full-length
100 μL
Human
LOC653107 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.08kDa
Transient overexpression cell lysate was tested with anti-LOC653107 antibody (Mfr. No. H00653145-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot