Learn More
Aviva Systems Biology Corp ZNF705D Antibody - middle region (ARP50437_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC728957. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC728957. Alternate names: ZNF705CApplicationWB. Species reactivity: Human
Catalog No. 50-014-60341