Learn More
Aviva Systems Biology Corp LOC729185 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP49228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC729185. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOC729185. Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-60046
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.