Learn More
Aviva Systems Biology Corp LOC729745 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP50966_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC729745. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOC729745. Alternate names: -ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-60476
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.