Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LOC81691 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559333
Description
Immunogen sequence: ALNILTGKDW KLKGRHALTP RHLHAWLRGL PPESTRLPGL RVVPPPFEQE ALQTLKLDHP KIAAWRWSRK IGKLYNSLCP GTLCLILLPG TKSTHGSLSG LGLMGI Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
REXO5 (RNA Exonuclease 5) is a Protein Coding gene. Among its related pathways are Ribosome biogenesis in eukaryotes. An important paralog of this gene is REXO1.
Specifications
LOC81691 | |
Polyclonal | |
Unconjugated | |
REXO5 | |
44M2.3; Exonuclease NEF-sp; LOC81691; putative RNA exonuclease NEF-sp; REXO5; RNA exonuclease 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96IC2 | |
REXO5 | |
Recombinant protein corresponding to Human LOC81691. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction