Learn More
Poltex Locking N95 Mask Dispenser
Holds and dispenses one box of N95 Masks.
$138.40 - $154.30
Specifications
For Use With (Application) Healthcare
For Use With (Equipment) N95 masks
Material PETG
Product Type Locking N95 Dispenser
View More Specs
Alert:
Consumables shown for demonstration purposes only and are not included with the product.
Products 3
Catalog Number Mfr. No. Mounting Price Quantity & Availability  
Catalog Number Mfr. No. Mounting Price Quantity & Availability  
22-000-462
View Documents
Poltex
N95MSKLCKW
Wall
Each for $138.40
Add to cart
22-000-463
View Documents
Poltex
N95MSKLCKT
VHB Tape
Each for $152.00
Add to cart
22-000-464
View Documents
Poltex
N95MSKLCKM
Magnet
Each for $154.30
Add to cart
Description
Description
  • Made with extremely durable, break resistant 3/16 in. thick PETG
  • Choose from keyhole slots for wall mounting, or powerful magnets.
  • Locking, sliding lid keeps dust out
  • Hardware not included for wall mount unit (-W). Customer must provide their own screw/anchor for their wall type.
Specifications
Specifications
Healthcare
N95 masks
PETG
Locking N95 Dispenser