Learn More
Brady™ Lockout Center Board
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Lockout Center Board is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ 3044
Catalog No. 19-103-998
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Board
Yellow
Acrylic
14 in. x 20 in.
Safety
20 in.
14 in.