Learn More
Brady™ Lockout Tagout Kit, Option #3
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Lockout Tagout Kit, Option #3 is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Supplier:  Brady™ LK329E
Catalog No. 19-104-219
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
Mechanical Risk Lockouts
Powder-coated Metal
1 Kit
Red
(3) yellow lockout hasp; (2) 4 in x 6 in prinzing magnetic danger sign; (1) prinzing osha summary sheet; (3) red padlock; (5) prinzing danger tag; (1) ac sensor; (1) pad of prinzing lockout tagout checklists; (1) prinzing lockout keychain; (1) prinzing red metal tool box; (1) prinzing tool box divider-lab safety
Product Suggestions
Product Suggestions