Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Lockout/Tagout: Your Key to Safety Training Program Accessory - Employee Booklet, English

Catalog No. 19103318 Shop All Brady Corporation Products
Click to view available options
Width (Metric):
133.35 mm

Use this extra employee handbook with the Lockout/Tagout: Your Key to Safety Program to train employees to protect themselves during machine servicing and maintenance.

  • This extra employee handbook is designed for use with the Lockout/Tagout: Your Key to Safety Program
  • Shows employees how to recognize all potential hazards and how to handle them safely
  • Package includes employee handbook only (full training program sold separately)
  • Helps employees and businesses comply with OSHA's Lockout/Tagout Standard (29 CFR, Part 1910.147)

Specifications

Format Booklet
Width (Metric) 133.35 mm
Quantity Each
For Use With (Application) Lockout Tagout Training
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.