Learn More
Loctite™ Super Bonder Adhesives
Instant adhesives with gap filling capability
Manufacturer:  Nasco 44243050
 View more versions of this product
Catalog No. 19-313-596
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
MIL-A-46050C
Cyanoacrylate
Bottle
-65° to 180°F
80cP
430 Super Bonder Adhesive
Clear
Used for bonding close fitting metal parts
Each
-53.88° to 82.22°C
1 oz.
29.57mL