Learn More
LOE 908 hydrochloride, Tocris
Broad spectrum cation channel blocker; neuroprotective
Manufacturer:  R&D Systems 2314/10
Quantity 10mg
 View more versions of this product
Catalog No. 23-141-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
749.29
LOE 908 hydrochloride
C41H48N2O9·HCl
3,4-Dihydro-6,7-dimethoxy-α-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]-1-isoquinolineacetamide hydrochloride
Specifications
10mg
>98%
749.29
LOE 908 hydrochloride
143482-60-4
C41H48N2O9·HCl
3,4-Dihydro-6,7-dimethoxy-α-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]-1-isoquinolineacetamide hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature