Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LOE 908 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Broad spectrum cation channel blocker. Inhibits several cation channels including store-operated calcium channels (SOCs), voltage-operated calcium channels (VOCs), non-selective cation channels (NSCCs), AMPA, NMDA, Na+ and K+ channels. Neuroprotective; reduces cortical infarct size and improves neurological outcome following middle cerebral artery occlusion.
Specifications
Quantity | 10 mg |
Formula Weight | 749.29 |
Percent Purity | >98% |
Chemical Name or Material | LOE 908 hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction