Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOE 908 hydrochloride, Tocris Bioscience™

Catalog Number 231450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
143482-60-4
C41H49ClN2O9
749.298
UDKSNBMNKLDHNK-UHFFFAOYSA-N
loe 908 hydrochloride, pinokalant hydrochloride, dsstox_cid_26305, dsstox_rid_81526, dsstox_gsid_46305, 2-6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl-2-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl acetamide hydrochloride, 3,4-dihydro-6,7-dimethoxy-?-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl-1-isoquinolineacetamide hydrochloride
56846451
2-(6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl)-2-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]acetamide;hydrochloride
COC1=C(C(=C(C=C1)CCN(CCC2=C(C(=C(C=C2)OC)OC)OC)C(=O)C(C3=CC=CC=C3)C4=NCCC5=CC(=C(C=C54)OC)OC)OC)OC.Cl
This item is not returnable. View return policy
143482-60-4
C41H49ClN2O9
749.298
UDKSNBMNKLDHNK-UHFFFAOYSA-N
loe 908 hydrochloride, pinokalant hydrochloride, dsstox_cid_26305, dsstox_rid_81526, dsstox_gsid_46305, 2-6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl-2-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl acetamide hydrochloride, 3,4-dihydro-6,7-dimethoxy-?-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl-1-isoquinolineacetamide hydrochloride
56846451
2-(6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl)-2-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]acetamide;hydrochloride
COC1=C(C(=C(C=C1)CCN(CCC2=C(C(=C(C=C2)OC)OC)OC)C(=O)C(C3=CC=CC=C3)C4=NCCC5=CC(=C(C=C54)OC)OC)OC)OC.Cl

Broad spectrum cation channel blocker; neuroprotective

Broad spectrum cation channel blocker. Inhibits several cation channels including store-operated calcium channels (SOCs), voltage-operated calcium channels (VOCs), non-selective cation channels (NSCCs), AMPA, NMDA, Na+ and K+ channels. Neuroprotective; reduces cortical infarct size and improves neurological outcome following middle cerebral artery occlusion.
Quantity 50 mg
Formula Weight 749.29
Percent Purity >98%
Chemical Name or Material LOE 908 hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit