Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lofepramine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Serotonin and noradrenalin re-uptake inhibitor (SNRI) that is metabolized to desipramine. Produces inhibition of liver tryptophan pyrollase activity in vitro and displays antidepressant properties in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 418.97 |
Percent Purity | >99% |
Chemical Name or Material | Lofepramine |
Safety and Handling
Recommended Storage : Store at +4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title