Learn More
MilliporeSigma™ Loffler's Methylene Blue Solution
Used as bacteriological counter staining of gonococcal, lactic acid bacteria.
$31.57 - $201.71
Specifications
Storage Requirements 15° to 25°C
View More Specs
Products 3
Catalog Number Mfr. No. Packaging Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Type Quantity Price Quantity & Availability  
M1012872500 View Documents MilliporeSigma™
1.01287.2500
Plastic Bottle 2.5L Each for $201.71
Add to cart
M1012870100 View Documents MilliporeSigma™
1.01287.0100
Glass Bottle 100mL Each for $31.57
Add to cart
M1012870500 View Documents MilliporeSigma™
1.01287.0500
Glass Bottle 500mL Each for $50.31
Add to cart
Description
Description
  • Visualizes pole corpuscles of pasteurella
  • Used in combination with Ziehl-Neelsen carbol fuchsin solution hydrochloric acid in ethanol to stain mycobacteria
Specifications
Specifications
15° to 25°C