Learn More
Labconco™ Logic Vue Canopy Connection Kit
SDP
Logic Vue Canopy Connection for thimble exhausting Logic Vue Class II Enclosures. Canopy maintains a negative pressure connection to house exhaust. Detects insufficient exhaust in less than one second and signals Logic Vue LCD to show an alarm.
Supplier:  Labconco™ 3347801
Catalog No. 10-304-208
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Ventus Logic Vue Canopy Connection Kit, 4 ft.: For thimble ducting Logic Vue Class II Enclosures.
  • Energy-conserving design maintains safe negative pressure exhaust flow.
  • The built-in sensor recognizes building exhaust failure in less than one second and sends an alarm signal to trigger audio and visual alarms via the LCD screen.
  • Adds 9.8 in. to overall depth of Logic Vue.
Specifications
Specifications
Canopy Connection Kit
Product Suggestions
Product Suggestions