Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VWA5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550363
Description
The antibody detects endogenous levels of total LOH11CR2A protein.
VWA5A Polyclonal antibody specifically detects VWA5A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
VWA5A | |
Polyclonal | |
Unconjugated | |
Vwa5a | |
5830475I06Rik; AW552491; BCSC1; BCSC-1; breast cancer suppressor candidate 1; E130119J22; Loh11cr2a; loss of heterozygosity 11 chromosomal region 2 gene A protein; Loss of heterozygosity 11 chromosomal region 2 gene A protein homolog; loss of heterozygosity, 11, chromosomal region 2, gene A; loss of heterozygosity, 11, chromosomal region 2, gene A homolog; Masa1; masa-1; Mast cell surface antigen 1; mast cell surface antigen-1; ortholog of mouse AW551984; von Willebrand factor A domain containing 5A; von Willebrand factor A domain-containing protein 5A; VWA5A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4013 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O00534 | |
Vwa5a | |
Fusion protein corresponding to residues near the C terminal of human von Willebrand factor A domain containing 5A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction