Learn More
LOK Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300400AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Serine/threonine-protein kinase involved in regulation of lymphocyte migration. Phosphorylates MSN, and possibly PLK1. Involved in regulation of lymphocyte migration by mediating phosphorylation of ERM proteins such as MSN. Acts as a negative regulator of MAP3K1/MEKK1. May also act as a cell cycle regulator by acting as a polo kinase kinase: mediates phosphorylation of PLK1 in vitro; however such data require additional evidences in vivo.Specifications
| LOK | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O94804 | |
| STK10 | |
| Between 900 and C-term | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| STK10 | |
| Serine/threonine protein kinase 10; Lymphocyte-oriented kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6793 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.