Brady™ Long Body Keyed Alike Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
Manufacturer: Brady™ 123431
 View more versions of this product
Catalog No. 19-103-514
  Check Availability
Add to Cart
    Specifications
    Color White
    Key Type Alike
    Material Nylon plastic
    Quantity 6/Pk.
  • Description & Specifications
Related Products
Product Content Feedback