Learn More
Brady™ Long Body Keyed-Different Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks electrical current from passing padlock shackle to cylinder
Manufacturer:  Brady™ 123408
 View more versions of this product
Catalog No. 19-103-491
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Each
Yellow
Different
Nylon plastic