Learn More
Lonza™ Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions
A selection of concentrated and premix solutions for automated, high accuracy DNA sequencing applications. Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions offer greater flexibility to optimize run conditions and change gel percentages.
Manufacturer:  Lonza™ 50611
 View more versions of this product
Catalog No. BMA50611
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Concentrated Solutions 
  • Offer more flexibility to optimize run conditions and change gel percentages
  • Require the addition of urea, buffer, APS, and TEMED
Premix Solutions
  • Contain all gel components for increased run-to-run consistency and ease of use
  • Require the addition of APS and TEMED for gel polymerization
Specifications
Specifications
Liquid
DNA and RNA Sequencing, genotyping
250 mL
Agarose Electrophoresis Gels
Agarose
18° to 26°C