Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions

A selection of concentrated and premix solutions for automated, high accuracy DNA sequencing applications. Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions offer greater flexibility to optimize run conditions and change gel percentages.

Specifications

Format Liquid
Product Type Specs Agarose Electrophoresis Gels
For Use With (Application) DNA and RNA Sequencing, genotyping
Material Agarose
Storage Requirements 18° to 26°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA50615
View Documents
Lonza™
50615
1 L This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
BMA50611
View Documents
Lonza™
50611
250 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Concentrated Solutions

  • Offer more flexibility to optimize run conditions and change gel percentages
  • Require the addition of urea, buffer, APS, and TEMED

Premix Solutions

  • Contain all gel components for increased run-to-run consistency and ease of use
  • Require the addition of APS and TEMED for gel polymerization
Specifications

Specifications

Liquid
DNA and RNA Sequencing, genotyping
18° to 26°C
Agarose Electrophoresis Gels
Agarose
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit